Assay Details
Gene Symbol: pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105132.2 | NP_001098602.1 | 6 - 7 | 1037 | 83 | |
GenBank mRNA | BC152623.1 | - | 6 - 7 | 741 | 83 | |
BC154028.1 | - | 6 - 7 | 798 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
