Assay Details
Gene Symbol: mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199716.1 | NP_956010.1 | 2 - 3 | 717 | 121 | |
GenBank mRNA | BC049527.1 | - | 2 - 3 | 717 | 121 | |
BC067183.1 | - | 2 - 3 | 551 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
