Assay Details
Gene Symbol: gabra6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200731.1 | NP_957025.1 | 9 - 10 | 1253 | 133 | |
XM_005173112.4 | XP_005173169.1 | 9 - 10 | 1772 | 133 | ||
GenBank mRNA | BC059508.1 | - | 9 - 10 | 1253 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
