Assay Details
Gene Symbol: lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002606.1 | NP_001002606.1 | 4 - 5 | 592 | 60 | |
XM_005162061.4 | XP_005162118.1 | 4 - 5 | 519 | 60 | ||
XM_017353544.2 | XP_017209033.1 | 4 - 5 | 669 | 60 | ||
GenBank mRNA | BC075986.1 | - | 4 - 5 | 592 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
