Assay Details
Gene Symbol: lemd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002606.1 | NP_001002606.1 | 5 - 6 | 654 | 77 | |
XM_005162053.4 | XP_005162110.1 | 9 - 10 | 1096 | 77 | ||
XM_005162054.4 | XP_005162111.1 | 9 - 10 | 1074 | 77 | ||
XM_005162055.4 | XP_005162112.1 | 8 - 9 | 1026 | 77 | ||
XM_005162058.4 | XP_005162115.1 | 6 - 7 | 837 | 77 | ||
XM_005162059.4 | XP_005162116.1 | 6 - 7 | 567 | 77 | ||
XM_005162060.4 | XP_005162117.1 | 6 - 7 | 674 | 77 | ||
XM_005162061.4 | XP_005162118.1 | 5 - 6 | 581 | 77 | ||
XM_009296725.3 | XP_009295000.1 | 7 - 8 | 935 | 77 | ||
XM_017353540.2 | XP_017209029.1 | 9 - 10 | 1079 | 77 | ||
XM_017353541.2 | XP_017209030.1 | 8 - 9 | 1008 | 77 | ||
XM_017353542.2 | XP_017209031.1 | 9 - 10 | 807 | 77 | ||
XM_017353543.2 | XP_017209032.1 | 9 - 10 | 988 | 77 | ||
XM_017353544.2 | XP_017209033.1 | 5 - 6 | 731 | 77 | ||
XM_021469779.1 | XP_021325454.1 | 10 - 11 | 1162 | 77 | ||
XM_021469780.1 | XP_021325455.1 | 7 - 8 | 918 | 77 | ||
XM_021469781.1 | XP_021325456.1 | 7 - 8 | 913 | 77 | ||
XM_021469782.1 | XP_021325457.1 | 6 - 7 | 820 | 77 | ||
GenBank mRNA | BC075986.1 | - | 5 - 6 | 654 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
