Assay Details
Gene Symbol: gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004018.1 | NP_001004018.1 | 2 - 3 | 487 | 87 | |
XM_005163776.4 | XP_005163833.1 | 3 - 4 | 778 | 87 | ||
GenBank mRNA | AY690668.1 | - | 2 - 3 | 487 | 87 | |
AY989813.1 | - | 2 - 3 | 487 | 87 | ||
BC163671.1 | - | 2 - 3 | 531 | 87 | ||
BC163674.1 | - | 2 - 3 | 531 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
