Assay Details
Gene Symbol: smarcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007296.1 | NP_001007297.1 | 1 - 2 | 139 | 121 | |
GenBank mRNA | BC085668.1 | - | 1 - 2 | 139 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
