Assay Details
Gene Symbol: bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012245.1 | NP_001012245.1 | 1 - 2 | 122 | 91 | |
XM_005172968.4 | XP_005173025.1 | 1 - 2 | 395 | 91 | ||
GenBank mRNA | AY424950.1 | - | 1 - 2 | 122 | 91 | |
BC109452.1 | - | 1 - 2 | 339 | 91 | ||
BC171679.1 | - | 1 - 2 | 86 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
