Assay Details
Gene Symbol: cog8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013506.1 | NP_001013524.1 | 2 - 3 | 549 | 137 | |
GenBank mRNA | BC090428.1 | - | 2 - 3 | 549 | 137 | |
BC165858.1 | - | 2 - 3 | 569 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
