Assay Details
Gene Symbol: cog8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013506.1 | NP_001013524.1 | 3 - 4 | 1371 | 72 | |
GenBank mRNA | BC090428.1 | - | 3 - 4 | 1371 | 72 | |
BC165858.1 | - | 3 - 4 | 1391 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
