Assay Details
Gene Symbol: gabrb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024387.2 | NP_001019558.2 | 6 - 7 | 745 | 126 | |
XM_005173103.3 | XP_005173160.1 | 6 - 7 | 695 | 126 | ||
GenBank mRNA | AJ302649.1 | - | 832 | 126 | ||
BC115078.1 | - | 6 - 7 | 711 | 126 | ||
EE690598.1 | - | 4 - 5 | 520 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
