Assay Details
Gene Symbol: gabrb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024387.2 | NP_001019558.2 | 9 - 10 | 1280 | 64 | |
GenBank mRNA | AJ302649.1 | - | 1367 | 64 | ||
BC115078.1 | - | 9 - 10 | 1246 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
