Assay Details
Gene Symbol: olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039932.1 | NP_001035021.1 | 5 - 6 | 849 | 114 | |
XM_005159455.3 | XP_005159512.1 | 5 - 6 | 631 | 114 | ||
XM_021468123.1 | XP_021323798.1 | 5 - 6 | 853 | 114 | ||
GenBank mRNA | BC114310.1 | - | 5 - 6 | 849 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
