Assay Details
Gene Symbol: srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045427.1 | NP_001038892.1 | 2 - 3 | 365 | 142 | |
XM_005172446.4 | XP_005172503.1 | 2 - 3 | 385 | 142 | ||
XM_017357373.2 | XP_017212862.1 | 2 - 3 | 492 | 142 | ||
XM_017357374.2 | XP_017212863.1 | 2 - 3 | 386 | 142 | ||
GenBank mRNA | BC122414.1 | - | 2 - 3 | 365 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
