Assay Details
Gene Symbol: LOC556326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099446.1 | NP_001092916.1 | 7 - 8 | 864 | 114 | |
GenBank mRNA | CU458952.1 | - | 7 - 8 | 864 | 114 |
Gene Symbol: il11ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113499.1 | NP_001106971.1 | 7 - 8 | 820 | 114 | |
XM_009305183.3 | XP_009303458.1 | 7 - 8 | 894 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
