Assay Details
Gene Symbol: naxd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110120.1 | NP_001103590.1 | 5 - 6 | 793 | 65 | |
NM_001328150.1 | NP_001315079.1 | 5 - 6 | 1783 | 65 | ||
XM_005167456.4 | XP_005167513.1 | 5 - 6 | 1762 | 65 | ||
XM_005167457.4 | XP_005167514.1 | 5 - 6 | 1744 | 65 | ||
XM_005167459.2 | XP_005167516.1 | 5 - 6 | 851 | 65 | ||
XM_005167460.4 | XP_005167517.1 | 5 - 6 | 1129 | 65 | ||
XM_005167462.4 | XP_005167519.1 | 5 - 6 | 1096 | 65 | ||
XM_005167463.4 | XP_005167520.1 | 4 - 5 | 812 | 65 | ||
XM_005167464.4 | XP_005167521.1 | 4 - 5 | 667 | 65 | ||
GenBank mRNA | BC153915.1 | - | 5 - 6 | 793 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
