Assay Details
Gene Symbol: ntn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130998.1 | NP_571073.1 | 5 - 6 | 1485 | 85 | |
XM_005171551.4 | XP_005171608.1 | 6 - 7 | 1774 | 85 | ||
XM_005171552.4 | XP_005171609.1 | 6 - 7 | 1769 | 85 | ||
GenBank mRNA | AF017420.1 | - | 5 - 6 | 1485 | 85 | |
BC163393.1 | - | 5 - 6 | 1512 | 85 | ||
CU638800.2 | - | 5 - 6 | 1485 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
