Assay Details
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 50 - 51 | 3659 | 102 | |
XM_005166863.4 | XP_005166920.1 | 49 - 50 | 3591 | 102 | ||
GenBank mRNA | BC163934.1 | - | 50 - 51 | 3651 | 102 | |
BC167477.1 | - | 49 - 50 | 3555 | 102 | ||
BC168504.1 | - | 50 - 51 | 3740 | 102 | ||
DQ335127.1 | - | 50 - 51 | 3659 | 102 | ||
U23822.1 | - | 7 - 8 | 534 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
