Assay Details
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 7 - 8 | 588 | 68 | |
XM_005166863.4 | XP_005166920.1 | 6 - 7 | 520 | 68 | ||
GenBank mRNA | BC163934.1 | - | 7 - 8 | 580 | 68 | |
BC167477.1 | - | 6 - 7 | 484 | 68 | ||
BC168504.1 | - | 7 - 8 | 669 | 68 | ||
DQ335127.1 | - | 7 - 8 | 588 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
