Assay Details
Gene Symbol: klhl24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200440.1 | NP_956734.1 | 4 - 5 | 1342 | 81 | |
GenBank mRNA | BC054921.1 | - | 4 - 5 | 1342 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
