Assay Details
Gene Symbol: cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205700.1 | NP_991263.1 | 2 - 3 | 382 | 143 | |
GenBank mRNA | BC065947.1 | - | 2 - 3 | 382 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
