Assay Details
Gene Symbol: tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207100.2 | NP_996983.1 | 675 | 105 | ||
GenBank mRNA | BC066516.1 | - | 2 - 3 | 243 | 105 | |
BC124418.1 | - | 675 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
