Assay Details
Gene Symbol: rtkn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328566.1 | NP_001315495.1 | 4 - 5 | 630 | 123 | |
GenBank mRNA | BC081383.1 | - | 4 - 5 | 676 | 123 | |
EH585343.1 | - | 4 - 5 | 626 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
