Assay Details
Gene Symbol: gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008731.1 | NP_001008731.1 | 2 - 3 | 493 | 75 | |
GenBank mRNA | AY770492.1 | - | 2 - 3 | 493 | 75 | |
BC163268.1 | - | 2 - 3 | 517 | 75 | ||
BC163279.1 | - | 2 - 3 | 517 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
