Assay Details
Gene Symbol: nmnat1-rbp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017629.2 | NP_001017629.1 | 3 - 4 | 339 | 82 | |
GenBank mRNA | BC092821.1 | - | 3 - 4 | 341 | 82 | |
BC165446.1 | - | 2 - 3 | 331 | 82 | ||
CO804002.1 | - | 3 - 4 | 349 | 82 | ||
CT621800.2 | - | 2 - 3 | 322 | 82 |
Gene Symbol: nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328328.1 | NP_001315257.1 | 3 - 4 | 354 | 82 | |
GenBank mRNA | CK016190.1 | - | 2 - 3 | 309 | 82 | |
EH495187.1 | - | 3 - 4 | 380 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
