Assay Details
Gene Symbol: emc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025515.2 | NP_001020686.1 | 1 - 2 | 353 | 144 | |
GenBank mRNA | BC116477.1 | - | 1 - 2 | 353 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
