Assay Details
Gene Symbol: psmb13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131677.2 | NP_571752.2 | 3 - 4 | 304 | 63 | |
GenBank mRNA | AF155577.1 | - | 3 - 4 | 256 | 63 | |
BC092899.1 | - | 3 - 4 | 285 | 63 | ||
BC165548.1 | - | 3 - 4 | 294 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
