Assay Details
Gene Symbol: psmb13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131677.2 | NP_571752.2 | 6 - 7 | 620 | 143 | |
GenBank mRNA | AF155577.1 | - | 6 - 7 | 572 | 143 | |
BC092899.1 | - | 6 - 7 | 143 | |||
BC165548.1 | - | 6 - 7 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
