Assay Details
Gene Symbol: klhl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199783.1 | NP_956077.1 | 1 - 2 | 562 | 106 | |
GenBank mRNA | BC045881.1 | - | 1 - 2 | 562 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
