Assay Details
Gene Symbol: pfn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201466.2 | NP_958874.2 | 1 - 2 | 295 | 98 | |
XM_005171152.2 | XP_005171209.1 | 1 - 2 | 365 | 98 | ||
GenBank mRNA | BC045843.1 | - | 1 - 2 | 264 | 98 | |
BC078650.1 | - | 1 - 2 | 295 | 98 | ||
BC164667.1 | - | 1 - 2 | 165 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
