Assay Details
Gene Symbol: gamt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105595.1 | NP_001099065.1 | 5 - 6 | 604 | 139 | |
GenBank mRNA | AF425745.1 | - | 5 - 6 | 627 | 139 | |
BC107607.1 | - | 5 - 6 | 651 | 139 | ||
BC153601.1 | - | 5 - 6 | 604 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
