Assay Details
Gene Symbol: adra2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207639.1 | NP_997522.1 | 1 | 1054 | 113 | |
GenBank mRNA | BC163670.1 | - | 1 | 1096 | 113 | |
BC163673.1 | - | 1 | 1096 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
