Assay Details
Gene Symbol: nkx1.2la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212830.2 | NP_997995.2 | 1 - 2 | 322 | 60 | |
GenBank mRNA | AB067732.1 | - | 1 - 2 | 292 | 60 | |
BC076418.1 | - | 1 - 2 | 326 | 60 | ||
CT721252.2 | - | 1 - 2 | 304 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC076418.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
