Assay Details
Gene Symbol: ppp3r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004553.1 | NP_001004553.1 | 4 - 5 | 470 | 61 | |
XM_005160126.4 | XP_005160183.1 | 3 - 4 | 333 | 61 | ||
GenBank mRNA | BC081617.1 | - | 4 - 5 | 470 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
