Assay Details
Gene Symbol: mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009297359.3 | XP_009295634.2 | 4 - 5 | 558 | 69 | |
XM_009297362.3 | XP_009295637.2 | 4 - 5 | 557 | 69 | ||
XM_017353906.2 | XP_017209395.2 | 5 - 6 | 684 | 69 | ||
XM_017353907.2 | XP_017209396.2 | 5 - 6 | 684 | 69 | ||
XM_017353908.2 | XP_017209397.2 | 5 - 6 | 684 | 69 | ||
XM_017353909.2 | XP_017209398.2 | 5 - 6 | 684 | 69 | ||
XM_017353910.2 | XP_017209399.2 | 5 - 6 | 684 | 69 | ||
XM_017353911.2 | XP_017209400.2 | 5 - 6 | 684 | 69 | ||
XM_017353912.2 | XP_017209401.2 | 5 - 6 | 684 | 69 | ||
XM_017353913.2 | XP_017209402.2 | 5 - 6 | 684 | 69 | ||
XM_017353914.2 | XP_017209403.2 | 5 - 6 | 684 | 69 | ||
XM_021470301.1 | XP_021325976.1 | 5 - 6 | 684 | 69 | ||
GenBank mRNA | BC085650.1 | - | 3 - 4 | 442 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
