Assay Details
Gene Symbol: elp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014322.2 | NP_001014344.2 | 13 - 14 | 1614 | 59 | |
GenBank mRNA | BC091787.1 | - | 11 - 12 | 1389 | 59 | |
BC096810.1 | - | 13 - 14 | 1614 | 59 | ||
BC165402.1 | - | 13 - 14 | 1520 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
