Assay Details
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 12 - 13 | 1686 | 73 | |
XM_005161411.4 | XP_005161468.1 | 11 - 12 | 73 | |||
GenBank mRNA | BC091956.1 | - | 12 - 13 | 1690 | 73 | |
BC165859.1 | - | 12 - 13 | 1453 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
