Assay Details
Gene Symbol: taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042744.2 | NP_001036209.1 | 1 - 2 | 170 | 66 | |
GenBank mRNA | BC098542.1 | - | 1 - 2 | 282 | 66 | |
BC116480.1 | - | 1 - 2 | 282 | 66 | ||
BC133110.1 | - | 1 - 2 | 282 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
