Assay Details
Gene Symbol: stag2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100028.1 | NP_001093498.1 | 29 - 30 | 3438 | 60 | |
XM_009301356.3 | XP_009299631.1 | 29 - 30 | 3499 | 60 | ||
XM_017356146.2 | XP_017211635.1 | 29 - 30 | 3581 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
