Assay Details
Gene Symbol: cx43.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131069.2 | NP_571144.2 | 2 - 3 | 1318 | 114 | |
GenBank mRNA | BC044357.1 | - | 2 - 3 | 1318 | 114 | |
L46801.1 | - | 2 - 3 | 1214 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
