Assay Details
Gene Symbol: cyp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131879.2 | NP_571954.1 | 3 - 4 | 1056 | 93 | |
GenBank mRNA | AB078927.1 | - | 3 - 4 | 1056 | 93 | |
AF210727.2 | - | 3 - 4 | 1031 | 93 | ||
AY398333.1 | - | 3 - 4 | 1030 | 93 | ||
BC094977.1 | - | 3 - 4 | 1056 | 93 | ||
BC165188.1 | - | 2 - 3 | 1001 | 93 | ||
CF997194.1 | - | 2 - 3 | 190 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
