Assay Details
Gene Symbol: cyp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131879.2 | NP_571954.1 | 5 - 6 | 1267 | 82 | |
GenBank mRNA | AB078927.1 | - | 5 - 6 | 1267 | 82 | |
AF210727.2 | - | 5 - 6 | 1242 | 82 | ||
AY398333.1 | - | 5 - 6 | 1241 | 82 | ||
CF997194.1 | - | 4 - 5 | 401 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
