Assay Details
Gene Symbol: stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173244.2 | NP_775351.2 | 8 - 9 | 1281 | 84 | |
XM_017353103.2 | XP_017208592.1 | 3 - 4 | 272 | 84 | ||
GenBank mRNA | AY099525.1 | - | 8 - 9 | 84 | ||
BC091991.1 | - | 8 - 9 | 1287 | 84 | ||
BC164583.1 | - | 7 - 8 | 939 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
