Assay Details
Gene Symbol: pex26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199920.1 | NP_956214.1 | 4 - 5 | 940 | 119 | |
GenBank mRNA | BC053199.1 | - | 4 - 5 | 940 | 119 | |
BC164102.1 | - | 4 - 5 | 874 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
