Assay Details
Gene Symbol: chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200014.1 | NP_956308.1 | 3 - 4 | 387 | 63 | |
XM_009301289.3 | XP_009299564.1 | 3 - 4 | 424 | 63 | ||
GenBank mRNA | BC045934.1 | - | 3 - 4 | 387 | 63 | |
BC065462.1 | - | 3 - 4 | 370 | 63 | ||
BC067569.1 | - | 3 - 4 | 363 | 63 | ||
BC153497.1 | - | 3 - 4 | 391 | 63 | ||
BC165138.1 | - | 3 - 4 | 291 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
