Assay Details
Gene Symbol: chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200014.1 | NP_956308.1 | 4 - 5 | 464 | 65 | |
XM_009301289.3 | XP_009299564.1 | 4 - 5 | 501 | 65 | ||
GenBank mRNA | AY398326.1 | - | 4 - 5 | 470 | 65 | |
BC045934.1 | - | 4 - 5 | 464 | 65 | ||
BC065462.1 | - | 4 - 5 | 447 | 65 | ||
BC067569.1 | - | 4 - 5 | 440 | 65 | ||
BC153497.1 | - | 4 - 5 | 468 | 65 | ||
BC165138.1 | - | 4 - 5 | 368 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
