Assay Details
Gene Symbol: cbx8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205616.2 | NP_991179.2 | 1 - 2 | 123 | 66 | |
GenBank mRNA | AY179330.1 | - | 1 - 2 | 118 | 66 | |
AY398319.1 | - | 1 - 2 | 118 | 66 | ||
BC125904.1 | - | 1 - 2 | 123 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
