Assay Details
Gene Symbol: ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002065.1 | NP_001002065.1 | 2 - 3 | 316 | 128 | |
GenBank mRNA | BC071388.1 | - | 2 - 3 | 316 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
