Assay Details
Gene Symbol: sf3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002067.1 | NP_001002067.1 | 3 - 4 | 434 | 102 | |
GenBank mRNA | BC071385.1 | - | 3 - 4 | 434 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
