Assay Details
Gene Symbol: bag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003533.2 | NP_001003533.1 | 1 - 2 | 177 | 71 | |
GenBank mRNA | BC078249.1 | - | 1 - 2 | 181 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
